Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSSC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297658.1 | NP_001284587.1 | 4 | 977 | ||
NM_001297659.1 | NP_001284588.1 | 3 | 825 | |||
NM_001297660.1 | NP_001284589.1 | 4 | 809 | |||
NM_005706.3 | NP_005697.2 | 3 | 668 | |||
XM_006718118.2 | XP_006718181.1 | 3 | 1707 | |||
XM_011519830.2 | XP_011518132.1 | 4 | 919 | |||
GenBank mRNA | AF125568.1 | - | 2 | 358 | ||
BC006091.1 | - | 3 | 381 | 3677524 | ||
BI768923.1 | - | 4 | 702 | |||
BX384913.2 | - | 4 | 538 |