Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SRRM1 SIRNA Assays ›
Gene Symbol:
SRRM1
Gene Name:
serine and arginine repetitive matrix 1
Gene Aliases:
160-KD, POP101, SRM160
Chromosome Location:
Chr.1: 24643012 - 24673281 on Build GRCh38
UniGene ID:
Hs.18192
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS173503
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SRRM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001303448.1
NP_001290377.1
16
2532
NM_001303449.1
NP_001290378.1
16
2461
NM_005839.3
NP_005830.2
15
2496
XM_005245717.2
XP_005245774.1
16
2448
XM_005245718.2
XP_005245775.1
16
2445
XM_005245720.2
XP_005245777.1
16
2439
XM_005245721.2
XP_005245778.1
15
2400
XM_005245722.2
XP_005245779.1
16
2358
XM_011540478.2
XP_011538780.1
16
2462
XM_011540479.1
XP_011538781.1
16
4409
XM_011540480.1
XP_011538782.1
17
3717
XM_011540481.1
XP_011538783.1
17
2759
XM_011540482.1
XP_011538784.1
16
4403
XM_011540483.1
XP_011538785.1
16
4319
XM_017000011.1
XP_016855500.1
16
2459
XM_017000012.1
XP_016855501.1
16
2453
XM_017000013.1
XP_016855502.1
15
2420
XM_017000014.1
XP_016855503.1
15
2417
XM_017000015.1
XP_016855504.1
15
2414
XM_017000016.1
XP_016855505.1
16
2394
XM_017000017.1
XP_016855506.1
16
2369
XM_017000018.1
XP_016855507.1
15
2403
XM_017000019.1
XP_016855508.1
15
2397
XM_017000020.1
XP_016855509.1
16
2355
XM_017000021.1
XP_016855510.1
16
2352
XM_017000022.1
XP_016855511.1
15
2310
XM_017000023.1
XP_016855512.1
16
4406
XM_017000024.1
XP_016855513.1
16
4400
XM_017000025.1
XP_016855514.1
15
4186
XM_017000026.1
XP_016855515.1
16
2536
XM_017000027.1
XP_016855516.1
15
4364
XM_017000028.1
XP_016855517.1
15
4361
XM_017000029.1
XP_016855518.1
16
2711
XM_017000030.1
XP_016855519.1
16
4316
XM_017000031.1
XP_016855520.1
17
2666
XM_017000032.1
XP_016855521.1
16
4313
XM_017000033.1
XP_016855522.1
15
4277
GenBank mRNA
AF048977.1
-
15
2271
AF419855.1
-
15
2493
AK302498.1
-
16
2351
AK307568.1
-
13
2172
BC036187.1
-
15
2324
4827738
KU178512.1
-
15
2272
KU178513.1
-
16
2314
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
RNA splicing, via transesterification reactions
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
RNA splicing
mRNA 3'-end processing
DNA binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0