Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEPT9 SIRNA Assays ›
Gene Symbol:
SEPT9
Gene Name:
septin 9
Gene Aliases:
AF17q25, MSF, MSF1, NAPB, PNUTL4, SINT1, SeptD1
Chromosome Location:
Chr.17: 77281410 - 77500596 on Build GRCh38
UniGene ID:
Hs.440932
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS173896
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEPT9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113491.1
NP_001106963.1
4
924
NM_001113492.1
NP_001106964.1
4
1099
NM_001113493.1
NP_001106965.1
3
1085
NM_001113494.1
NP_001106966.1
3
1038
NM_001113495.1
NP_001106967.1
2
738
NM_001113496.1
NP_001106968.1
2
333
NM_001293695.1
NP_001280624.1
3
867
NM_001293696.1
NP_001280625.1
2
149
NM_001293697.1
NP_001280626.1
2
362
NM_001293698.1
NP_001280627.1
2
448
NM_006640.4
NP_006631.2
3
1556
XM_005256962.1
XP_005257019.1
2
206
XM_006721643.2
XP_006721706.1
5
1027
XM_006721644.1
XP_006721707.1
2
249
XM_011524204.1
XP_011522506.1
5
1124
XM_011524206.1
XP_011522508.1
3
844
XM_011524207.1
XP_011522509.1
3
857
XM_011524208.2
XP_011522510.1
2
115
XM_017024031.1
XP_016879520.1
3
456
XM_017024032.1
XP_016879521.1
2
455
GenBank mRNA
AB023208.1
-
3
1038
AF123052.1
-
3
1519
AF142408.1
-
3
1001
AF189712.2
-
5
1051
AF189713.3
-
4
894
AJ312319.1
-
3
1085
AJ312320.1
-
2
127
AJ312321.1
-
3
1556
AJ312322.1
-
3
1027
AK022493.1
-
2
115
AK056495.1
-
2
738
AK290368.1
-
4
1099
AK299828.1
-
2
362
AK300268.1
-
5
758
AK300270.1
-
3
826
AK303449.1
-
2
142
AK304143.1
-
3
939
AK316473.1
-
2
448
BC021192.2
-
3
779
3346534
BC054004.1
-
5
754
6146448
BC114550.1
-
2
681
BT007215.1
-
3
744
CN393537.1
-
2
141
DB148645.1
-
2
333
KU178538.1
-
2
99
KU178539.1
-
3
744
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cell cycle
protein heterooligomerization
cell division
cell-cell adhesion
positive regulation of nonmotile primary cilium assembly
GTPase activity
protein binding
GTP binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-x42j5:80/100.66.73.110:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0