Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM114A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317993.1 | NP_001304922.1 | 3 | 255 | ||
NM_001317994.1 | NP_001304923.1 | 2 | 772 | |||
NM_018691.3 | NP_061161.2 | 2 | 208 | |||
XM_005268360.4 | XP_005268417.1 | 3 | 333 | |||
XM_005268361.4 | XP_005268418.1 | 2 | 236 | |||
XM_011537540.2 | XP_011535842.1 | 2 | 562 | |||
XM_011537541.2 | XP_011535843.1 | 2 | 1144 | |||
XM_011537542.2 | XP_011535844.1 | 2 | 433 | |||
XM_017008967.1 | XP_016864456.1 | 2 | 227 | |||
XM_017008968.1 | XP_016864457.1 | 2 | 249 | |||
GenBank mRNA | AF159700.1 | - | 2 | 701 | ||
AK024664.1 | - | 2 | 173 | |||
AK095446.1 | - | 3 | 322 | |||
AK225821.1 | - | 2 | 191 | |||
AK303865.1 | - | 2 | 180 | |||
AK313331.1 | - | 2 | 180 | |||
BC011414.1 | - | 3 | 190 | 3855873 | ||
BC050701.1 | - | 2 | 169 | |||
DA351656.1 | - | 2 | 208 | |||
DB022316.1 | - | 3 | 316 | |||
DB028061.1 | - | 2 | 454 |