Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LILRB1 SIRNA Assays ›
Gene Symbol:
LILRB1
Gene Name:
leukocyte immunoglobulin like receptor B1
Gene Aliases:
CD85J, ILT-2, ILT2, LIR-1, LIR1, MIR-7, MIR7, PIR-B, PIRB
Chromosome Location:
Chr.19: 54616930 - 54637925 on Build GRCh38
UniGene ID:
Hs.667388
Species:
Human
siRNA ID
HSS173929
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LILRB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001081637.2
NP_001075106.2
6
964
NM_001081638.3
NP_001075107.2
6
1070
NM_001081639.3
NP_001075108.2
6
1070
NM_001278398.2
NP_001265327.2
6
964
NM_001278399.2
NP_001265328.2
6
964
NM_006669.6
NP_006660.4
7
1384
NR_103518.2
6
964
XM_011526331.2
XP_011524633.1
5
872
XM_011526332.2
XP_011524634.1
5
872
XM_011526335.2
XP_011524637.1
5
872
XM_011526336.2
XP_011524638.1
5
872
XM_011526338.2
XP_011524640.1
6
1090
XM_017026182.1
XP_016881671.1
5
872
XM_017026183.1
XP_016881672.1
5
872
XM_017026184.1
XP_016881673.1
5
872
XM_017026185.1
XP_016881674.1
5
871
XM_017026186.1
XP_016881675.1
6
1024
XM_017026187.1
XP_016881676.1
7
1375
XM_017026188.1
XP_016881677.1
6
1024
XM_017026189.1
XP_016881678.1
6
1024
XM_017026190.1
XP_016881679.1
7
1379
XM_017026191.1
XP_016881680.1
5
872
XM_017026192.1
XP_016881681.1
6
1084
GenBank mRNA
AF009005.1
-
5
799
AF009006.1
-
5
799
AF009007.1
-
5
799
AF009220.1
-
5
799
AF009221.1
-
5
799
AF283984.1
-
6
974
AF283985.1
-
6
904
AK223310.1
-
6
997
AK292028.1
-
7
1094
AK292156.1
-
7
1384
BC015731.1
-
7
1139
4878915
EU915608.1
-
5
799
U82279.1
-
5
799
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin receptor superfamily
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of T cell mediated cytotoxicity
positive regulation of defense response to virus by host
adaptive immune response
T cell proliferation involved in immune response
negative regulation of cytokine secretion involved in immune response
immune response-inhibiting cell surface receptor signaling pathway
Fc receptor mediated inhibitory signaling pathway
signal transduction
response to virus
positive regulation of gene expression
negative regulation of serotonin secretion
receptor internalization
interferon-gamma production
negative regulation of interferon-gamma production
negative regulation of mononuclear cell proliferation
negative regulation of interferon-beta secretion
negative regulation of T cell proliferation
negative regulation of tumor necrosis factor biosynthetic process
positive regulation of apoptotic process
negative regulation of interferon-gamma biosynthetic process
negative regulation of cell cycle
negative regulation of endocytosis
positive regulation of cytolysis
positive regulation of transcription from RNA polymerase II promoter
negative regulation of natural killer cell mediated cytotoxicity
negative regulation of alpha-beta T cell activation
regulation of immune response
defense response to virus
negative regulation of calcium ion transport
cellular response to lipopolysaccharide
interferon-gamma secretion
dendritic cell differentiation
negative regulation of dendritic cell apoptotic process
negative regulation of interleukin-10 secretion
negative regulation of interleukin-12 secretion
negative regulation of CD8-positive, alpha-beta T cell activation
negative regulation of T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
positive regulation of gamma-delta T cell activation involved in immune response
negative regulation of dendritic cell differentiation
negative regulation of transforming growth factor-beta secretion
negative regulation of osteoclast development
protein phosphatase 1 binding
HLA-A specific inhibitory MHC class I receptor activity
HLA-B specific inhibitory MHC class I receptor activity
MHC class I receptor activity
SH2 domain binding
MHC class I protein binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-p89b7:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0