Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ?
Create Account
Applications
Real-Time PCR
Oligos, Primers, Probes and Genes
Cloning
Protein Biology
Cell Culture and Transfection
Cell and Gene Therapy
Cell Analysis
Mass Spectrometry
Chromatography
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Online Order
Custom Primers & TaqMan Probes
miRNA Mimics & Inhibitors
Stealth RNAi / siRNA
Silencer Select siRNAs
Custom DNA Oligos
GeneArt Services
GeneArt Strings DNA Fragments
TrueGuide CRISPR gRNA
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all online orderable products
Services
Custom Services
Instrument Qualification Services
Technical Services
Enterprise Services
Pipette Services
Sample Request
Instrument Maintenance Services
Repairs and Relocation Services
OEM and Licensing Services
Events, Seminars, Training
See all services
Support
Catalogs
Application Notes
Safety Data Sheets
Legal and Regulatory
Certificates of Analysis Search
Nunc/Nalgene Product Certificates
e-learning
FAQs
Learning Centers
Technical Support Centers
See all help and support topics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign In
Sign In
Don't have an account ?
Create Account
Connect Your Lab
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BRD8 SIRNA Assays ›
Gene Symbol:
BRD8
Gene Name:
bromodomain containing 8
Gene Aliases:
SMAP, SMAP2, p120
Chromosome Location:
Chr.5: 138139766 - 138178669 on Build GRCh38
UniGene ID:
Hs.519337
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS173959
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Contact Us
Your Price
Online offer:
3062.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BRD8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164326.1
NP_001157798.1
18
2415
NM_001300961.1
NP_001287890.1
18
2411
NM_001300962.1
NP_001287891.1
17
2336
NM_001300966.1
NP_001287895.1
19
2503
NM_006696.3
NP_006687.3
19
2625
NM_139199.1
NP_631938.1
18
2406
XM_005271855.2
XP_005271912.1
19
2616
XM_005271856.2
XP_005271913.1
19
2616
XM_005271857.2
XP_005271914.1
18
2406
XM_005271859.3
XP_005271916.1
17
2187
XM_005271860.3
XP_005271917.1
19
2616
XM_005271861.3
XP_005271918.1
18
2397
XM_011543109.1
XP_011541411.1
17
2454
XM_011543110.1
XP_011541412.1
19
2602
XM_017008969.1
XP_016864458.1
16
2235
XM_017008970.1
XP_016864459.1
17
2454
XM_017008971.1
XP_016864460.1
17
2454
XM_017008972.1
XP_016864461.1
18
2406
XM_017008973.1
XP_016864462.1
18
2397
XM_017008974.1
XP_016864463.1
16
2244
XM_017008975.1
XP_016864464.1
16
2235
XM_017008976.1
XP_016864465.1
17
2187
GenBank mRNA
AB209079.1
-
19
2571
AF016270.1
-
18
2557
AK289951.1
-
19
2585
AK293621.1
-
17
2296
AK297608.1
-
9
1694
AK297756.1
-
18
2371
AK312333.1
-
19
2585
AL136823.1
-
18
2379
BC008039.2
-
19
2567
3009915
BC008076.2
-
19
2567
3009915
X87613.1
-
18
2433
X94234.1
-
17
2345
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
signal transduction
cell surface receptor signaling pathway
intracellular receptor signaling pathway
regulation of growth
histone H4 acetylation
histone H2A acetylation
transcription factor activity, sequence-specific DNA binding
thyroid hormone receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0