Search
Search

B4GALT7| Gene Symbol: B4GALT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007255.2 | NP_009186.1 | 5 | 901 | ||
| XM_006714816.3 | XP_006714879.1 | 5 | 833 | |||
| XM_017008999.1 | XP_016864488.1 | 5 | 3024 | |||
| GenBank mRNA | AB028600.1 | - | 5 | 848 | ||
| AF142675.1 | - | 5 | 843 | |||
| AJ005382.1 | - | 5 | 808 | |||
| AK022566.1 | - | 3 | 1908 | |||
| AK023506.1 | - | 5 | 844 | |||
| AK095882.1 | - | 5 | 1505 | |||
| AK123092.1 | - | 1 | 1627 | |||
| AY358578.1 | - | 5 | 884 | |||
| BC007317.2 | - | 5 | 901 | 3049998 | ||
| BC062983.1 | - | 5 | 868 | 4577572 | ||
| BC072403.1 | - | 5 | 890 | 6094074 | ||