Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HELQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297755.1 | NP_001284684.1 | 14 | 2840 | ||
NM_001297756.1 | NP_001284685.1 | 15 | 3054 | |||
NM_001297757.1 | NP_001284686.1 | 14 | 2943 | |||
NM_133636.3 | NP_598375.2 | 15 | 3041 | |||
XM_005262711.1 | XP_005262768.1 | 15 | 2902 | |||
XM_005262713.2 | XP_005262770.1 | 15 | 3131 | |||
XM_017007679.1 | XP_016863168.1 | 14 | 2701 | |||
XM_017007680.1 | XP_016863169.1 | 14 | 2930 | |||
XM_017007682.1 | XP_016863171.1 | 15 | 3122 | |||
XM_017007683.1 | XP_016863172.1 | 13 | 2241 | |||
GenBank mRNA | AF436845.1 | - | 15 | 3002 | ||
AK308395.1 | - | 14 | 2781 | |||
AK312725.1 | - | 15 | 3018 | |||
AL512702.1 | - | 13 | 1705 | |||
BC111502.1 | - | 14 | 2863 | |||
CK905810.1 | - | 4 | 563 |