Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TANGO2 SIRNA Assays ›
Gene Symbol:
TANGO2
Gene Name:
transport and golgi organization 2 homolog
Gene Aliases:
C22orf25, MECRCN
Chromosome Location:
Chr.22: 20017000 - 20067164 on Build GRCh38
UniGene ID:
Hs.474233
Species:
Human
siRNA ID
HSS174917
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TANGO2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001283106.2
NP_001270035.1
6
701
NM_001283116.2
NP_001270045.1
5
455
NM_001283129.2
NP_001270058.1
5
695
NM_001283148.2
NP_001270077.1
5
455
NM_001283154.2
NP_001270083.1
5
567
NM_001283199.2
NP_001270128.1
5
567
NM_001283215.2
NP_001270144.1
5
695
NM_001322141.1
NP_001309070.1
5
695
NM_001322142.1
NP_001309071.1
5
455
NM_001322143.1
NP_001309072.1
5
768
NM_001322144.1
NP_001309073.1
5
695
NM_001322146.1
NP_001309075.1
5
605
NM_001322148.1
NP_001309077.1
5
772
NM_001322149.1
NP_001309078.1
5
695
NM_001322160.1
NP_001309089.1
5
657
NM_152906.6
NP_690870.3
5
567
NR_136211.1
5
528
XM_011529863.1
XP_011528165.1
5
606
XM_011529865.1
XP_011528167.1
4
471
XM_017028577.1
XP_016884066.1
5
528
XM_017028578.1
XP_016884067.1
5
547
XM_017028579.1
XP_016884068.1
5
576
XM_017028580.1
XP_016884069.1
6
665
XM_017028581.1
XP_016884070.1
5
659
XM_017028582.1
XP_016884071.1
6
742
XM_017028583.1
XP_016884072.1
5
489
XM_017028584.1
XP_016884073.1
5
752
XM_017028587.1
XP_016884076.1
5
659
XM_017028588.1
XP_016884077.1
5
508
GenBank mRNA
AK289862.1
-
6
670
AK295210.1
-
4
581
AK297922.1
-
5
413
AK301366.1
-
5
654
AK316020.1
-
5
416
AK316056.1
-
5
675
AL713640.1
-
2
195
BC041339.1
-
5
530
5284126
BG685068.1
-
3
305
BI544307.1
-
5
530
BI911898.1
-
5
518
BM048105.1
-
36
BP303570.1
-
4
352
BU173566.1
-
5
487
CF552364.1
-
4
426
CR456355.1
-
4
382
CR627452.1
-
5
410
DB021425.1
-
3
245
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-2px8h:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0