Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172070.3 | NP_742067.3 | 14, 15 | 2152 | ||
XM_005246299.4 | XP_005246356.1 | 14, 15 | 2197 | |||
XM_005246300.4 | XP_005246357.1 | 14, 15 | 2152 | |||
XM_006712268.3 | XP_006712331.1 | 14, 15 | 2194 | |||
XM_006712269.3 | XP_006712332.1 | 14, 15 | 2152 | |||
XM_006712270.3 | XP_006712333.2 | 13, 14 | 2053 | |||
XM_006712271.3 | XP_006712334.1 | 14, 15 | 2193 | |||
XM_011510604.2 | XP_011508906.1 | 14, 15 | 2152 | |||
XM_017003356.1 | XP_016858845.1 | 14, 15 | 2152 | |||
XM_017003357.1 | XP_016858846.1 | 14, 15 | 2152 | |||
XM_017003358.1 | XP_016858847.1 | 14, 15 | 2152 | |||
XM_017003359.1 | XP_016858848.1 | 14, 15 | 2152 | |||
GenBank mRNA | DA352029.1 | - | 4, 5 | 350 |