Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173077.2 | NP_775100.1 | 5 | 454 | ||
XM_017003372.1 | XP_016858861.1 | 6 | 6561 | |||
GenBank mRNA | AJ422118.1 | - | 5 | 408 |