Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RALYL SIRNA Assays ›
Gene Symbol:
RALYL
Gene Name:
RALY RNA binding protein-like
Gene Aliases:
HNRPCL3
Chromosome Location:
Chr.8: 84182629 - 84921844 on Build GRCh38
UniGene ID:
Hs.121663
Species:
Human
siRNA ID
HSS175154
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RALYL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001100391.1
NP_001093861.1
2
339
NM_001100392.1
NP_001093862.1
3
582
NM_001100393.1
NP_001093863.1
2
550
NM_001287243.1
NP_001274172.1
2
1263
NM_173848.5
NP_776247.3
2
1263
XM_011517462.2
XP_011515764.1
3
1367
XM_017013058.1
XP_016868547.1
2
538
XM_017013059.1
XP_016868548.1
4
633
XM_017013060.1
XP_016868549.1
2
2783
XM_017013061.1
XP_016868550.1
2
1935
XM_017013062.1
XP_016868551.1
4
742
XM_017013063.1
XP_016868552.1
2
505
XM_017013064.1
XP_016868553.1
3
856
XM_017013065.1
XP_016868554.1
2
404
XM_017013066.1
XP_016868555.1
2
550
XM_017013067.1
XP_016868556.1
3
1441
XM_017013068.1
XP_016868557.1
4
633
XM_017013069.1
XP_016868558.1
4
689
XM_017013070.1
XP_016868559.1
3
533
XM_017013071.1
XP_016868560.1
2
404
XM_017013072.1
XP_016868561.1
2
491
XM_017013073.1
XP_016868562.1
2
491
XM_017013074.1
XP_016868563.1
2
404
XM_017013075.1
XP_016868564.1
2
489
XM_017013076.1
XP_016868565.1
2
284
XM_017013077.1
XP_016868566.1
3
1534
XM_017013078.1
XP_016868567.1
2
550
XM_017013079.1
XP_016868568.1
3
533
XM_017013080.1
XP_016868569.1
2
284
XM_017013081.1
XP_016868570.1
2
284
XM_017013082.1
XP_016868571.1
2
550
GenBank mRNA
AF495529.1
-
2
1263
AF495530.1
-
2
480
AK094543.1
-
2
494
AK095026.1
-
2
1079
AK095560.1
-
3
533
BC031090.1
-
2
328
5311935
DA143105.1
-
2
550
DN989439.1
-
2
344
Back To Top
More Information
Gene Ontology Categories:
Process(es)
nucleotide binding
protein binding
identical protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-2px8h:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0