Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CREM SIRNA Assays ›
Gene Symbol:
CREM
Gene Name:
cAMP responsive element modulator
Gene Aliases:
CREM-2, ICER, hCREM-2
Chromosome Location:
Chr.10: 35126841 - 35212958 on Build GRCh38
UniGene ID:
Hs.200250
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS175185
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
459.00
Your Price
Online offer:
459.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CREM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001267562.1
NP_001254491.1
3
542
NM_001267563.1
NP_001254492.1
2
401
NM_001267564.1
NP_001254493.1
2
313
NM_001267566.1
NP_001254495.1
2
167
NM_001881.3
NP_001872.3
3
525
NM_181571.2
NP_853549.1
4
381
NM_182769.2
NP_877570.1
2
167
NM_182770.2
NP_877571.1
2
167
NM_182771.1
NP_877572.1
2
313
NM_182772.1
NP_877573.1
2
313
NM_183011.1
NP_898829.1
3
270
NM_183012.1
NP_898830.1
3
270
NM_183013.2
NP_898831.1
4
623
NM_183060.2
NP_898883.1
3
283
XM_006717378.3
XP_006717441.1
4
363
XM_006717379.3
XP_006717442.1
4
414
XM_006717382.3
XP_006717445.1
4
355
XM_006717383.3
XP_006717446.1
4
355
XM_006717387.3
XP_006717450.1
4
355
XM_011519316.2
XP_011517618.1
5
502
XM_011519324.2
XP_011517626.1
4
355
XM_011519325.2
XP_011517627.1
4
437
XM_011519327.2
XP_011517629.1
3
335
XM_011519330.2
XP_011517632.1
2
278
XM_011519333.1
XP_011517635.1
2
173
XM_017015716.1
XP_016871205.1
3
474
XM_017015717.1
XP_016871206.1
3
401
XM_017015718.1
XP_016871207.1
4
355
XM_017015719.1
XP_016871208.1
3
355
XM_017015720.1
XP_016871209.1
4
355
XM_017015721.1
XP_016871210.1
3
344
XM_017015722.1
XP_016871211.1
3
257
XM_017015723.1
XP_016871212.1
3
414
XM_017015724.1
XP_016871213.1
4
626
XM_017015725.1
XP_016871214.1
2
130
XM_017015726.1
XP_016871215.1
3
282
XM_017015727.1
XP_016871216.1
2
282
XM_017015728.1
XP_016871217.1
4
626
XM_017015729.1
XP_016871218.1
2
268
XM_017015733.1
XP_016871222.1
2
153
XM_017015735.1
XP_016871224.1
2
130
XM_017015736.1
XP_016871225.1
2
158
XM_017015737.1
XP_016871226.1
2
282
GenBank mRNA
AK301906.1
-
2
132
AV707433.1
-
3
263
AY292864.1
-
2
158
BC017117.1
-
3
389
BC066342.1
-
3
443
BC090051.1
-
2
145
30387562
BG719264.1
-
2
318
BI459432.1
-
3
320
BU570307.1
-
2
149
CB996061.1
-
2
321
CD358637.1
-
2
261
S68134.1
-
3
315
S68271.1
-
3
315
Z15159.1
-
2
168
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
glucose metabolic process
transcription, DNA-templated
regulation of transcription, DNA-templated
fatty acid metabolic process
glycosphingolipid metabolic process
signal transduction
multicellular organism development
spermatogenesis
cell differentiation
circadian regulation of gene expression
regulation of circadian rhythm
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
retinoic acid receptor signaling pathway
response to cAMP
core promoter sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
cAMP response element binding protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-cqs7j:80/100.66.77.235:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0