Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CREM SIRNA Assays ›
Gene Symbol:
CREM
Gene Name:
cAMP responsive element modulator
Gene Aliases:
CREM-2, ICER, hCREM-2
Chromosome Location:
Chr.10: 35126841 - 35212958 on Build GRCh38
UniGene ID:
Hs.200250
Species:
Human
siRNA ID
HSS175186
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CREM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001267562.1
NP_001254491.1
3
576
NM_001267563.1
NP_001254492.1
2
435
NM_001267564.1
NP_001254493.1
2
347
NM_001267566.1
NP_001254495.1
2
201
NM_001881.3
NP_001872.3
3
559
NM_181571.2
NP_853549.1
4
415
NM_182769.2
NP_877570.1
2
201
NM_182770.2
NP_877571.1
2
201
NM_182771.1
NP_877572.1
2
347
NM_182772.1
NP_877573.1
2
347
NM_183011.1
NP_898829.1
3
304
NM_183012.1
NP_898830.1
3
304
NM_183013.2
NP_898831.1
4
657
NM_183060.2
NP_898883.1
3
317
XM_006717378.3
XP_006717441.1
4
397
XM_006717379.3
XP_006717442.1
4
448
XM_006717382.3
XP_006717445.1
4
389
XM_006717383.3
XP_006717446.1
4
389
XM_006717387.3
XP_006717450.1
4
389
XM_011519316.2
XP_011517618.1
5
536
XM_011519324.2
XP_011517626.1
4
389
XM_011519325.2
XP_011517627.1
4
471
XM_011519327.2
XP_011517629.1
3
369
XM_011519330.2
XP_011517632.1
2
312
XM_011519333.1
XP_011517635.1
2
207
XM_017015716.1
XP_016871205.1
3
508
XM_017015717.1
XP_016871206.1
3
435
XM_017015718.1
XP_016871207.1
4
389
XM_017015719.1
XP_016871208.1
3
389
XM_017015720.1
XP_016871209.1
4
389
XM_017015721.1
XP_016871210.1
3
378
XM_017015722.1
XP_016871211.1
3
291
XM_017015723.1
XP_016871212.1
3
448
XM_017015724.1
XP_016871213.1
4
660
XM_017015725.1
XP_016871214.1
2
164
XM_017015726.1
XP_016871215.1
3
316
XM_017015727.1
XP_016871216.1
2
316
XM_017015728.1
XP_016871217.1
4
660
XM_017015729.1
XP_016871218.1
2
302
XM_017015733.1
XP_016871222.1
2
187
XM_017015735.1
XP_016871224.1
2
164
XM_017015736.1
XP_016871225.1
2
192
XM_017015737.1
XP_016871226.1
2
316
GenBank mRNA
AK301906.1
-
2
166
AV707433.1
-
3
297
AY292864.1
-
2
192
BC017117.1
-
3
423
BC066342.1
-
3
477
BC090051.1
-
2
179
30387562
BG573680.1
-
3
528
BG719264.1
-
2
352
BI459432.1
-
3
354
BI561543.1
-
418
BU570307.1
-
2
183
CB996061.1
-
2
355
CD358637.1
-
2
295
D14825.1
-
4
336
S68134.1
-
3
349
S68271.1
-
3
349
Z15159.1
-
2
202
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
glucose metabolic process
transcription, DNA-templated
regulation of transcription, DNA-templated
fatty acid metabolic process
glycosphingolipid metabolic process
signal transduction
multicellular organism development
spermatogenesis
cell differentiation
circadian regulation of gene expression
regulation of circadian rhythm
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
retinoic acid receptor signaling pathway
response to cAMP
core promoter sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
cAMP response element binding protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-8dfkb:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0