Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CSNK2A1 SIRNA Assays ›
          Gene Symbol:
          CSNK2A1 CSNK2A3
          Gene Name:
          casein kinase 2 alpha 1
          casein kinase 2 alpha 3
          Gene Aliases:
          CK2A1, CKII, CSNK2A3
          CSNK2A1P
          Chromosome Location:
          Chr.20: 482694 - 543838 on Build GRCh38
          Chromosome Location:
          Chr.11: 11351942 - 11353357 on Build GRCh38
          UniGene ID:
          Hs.644056
          Hs.654675
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID HSS175396
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CSNK2A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001895.3 NP_001886.1 9, 10 997
          NM_177559.2 NP_808227.1 10, 11 1114
          NM_177560.2 NP_808228.1 8, 9 787
          GenBank mRNA AB451279.1 - 8, 9 721
          AB451411.1 - 8, 9 721
          AK302583.1 - 8, 9 676
          AK312535.1 - 10, 11 1005
          AW876962.1 - 3, 4 247
          BC011668.2 - 9, 10 869 4121882
          BC050036.1 - 9, 10 888 5298655
          BC053532.1 - 8, 9 816 3908058
          BC071167.1 - 8, 9 728 5772615
          BT019792.1 - 8, 9 721
          DQ354583.1 - 9, 10 863
          J02853.1 - 9, 10 874
          M55265.1 - 9, 10 869
          S53149.1 - 9, 10 874
          Gene Symbol: CSNK2A3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256686.1 NP_001243615.1 1 959

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_053824, XM_006235221, XM_006235220, L15618, BC091130.
          This siRNA also targets the following transcripts in Mouse: NM_007788, BC060742, U51866, U17112, AK158077, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein folding
          protein phosphorylation
          apoptotic process
          signal transduction
          positive regulation of cell proliferation
          Wnt signaling pathway
          positive regulation of Wnt signaling pathway
          positive regulation of cell growth
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of protein catabolic process
          protein autophosphorylation
          rhythmic process
          regulation of catalytic activity
          chaperone-mediated protein folding
          mitotic spindle checkpoint
          regulation of signal transduction by p53 class mediator
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          beta-catenin binding
          protein phosphatase regulator activity
          protein N-terminus binding
          Hsp90 protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline