Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100915.1 | NP_001094385.1 | 7 | 1140 | ||
GenBank mRNA | AK097481.1 | - | 7 | 1143 | ||
AK302567.1 | - | 6 | 2358 | |||
AK302748.1 | - | 7 | 1124 | |||
BC070103.1 | - | 7 | 2921 | |||
BC094790.1 | - | 6 | 2334 | |||
BC117335.1 | - | 7 | 1140 | 40125886 |