Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM98C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174905.3 | NP_777565.3 | 7, 8 | 931 | ||
GenBank mRNA | BC034383.1 | - | 5, 6 | 469 | ||
BC117258.1 | - | 7, 8 | 931 | 40125809 | ||
BC143830.1 | - | 7, 8 | 931 | 9052345 | ||
BC171747.1 | - | 4, 5 | 530 | |||
BI871449.1 | - | 4, 5 | 463 | |||
BX641942.1 | - | 1, 2 | 358 |