Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CTBP2 SIRNA Assays ›
          Gene Symbol:
          CTBP2
          Gene Name:
          C-terminal binding protein 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 124987849 - 125161061 on Build GRCh38
          UniGene ID:
          Hs.501345
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS175560
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CTBP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001083914.2 NP_001077383.1 7 1048
          NM_001290214.2 NP_001277143.1 7 1033
          NM_001290215.2 NP_001277144.1 7 1297
          NM_001321012.1 NP_001307941.1 7 992
          NM_001321013.1 NP_001307942.1 6 1193
          NM_001321014.1 NP_001307943.1 7 1294
          NM_001329.3 NP_001320.1 7 1036
          NM_022802.2 NP_073713.2 5 2353
          XM_005269561.2 XP_005269618.1 8 1219
          XM_005269564.2 XP_005269621.1 8 1427
          XM_005269567.2 XP_005269624.1 8 1426
          XM_005269568.4 XP_005269625.1 8 1154
          XM_005269569.2 XP_005269626.1 7 1115
          XM_005269571.2 XP_005269628.1 6 874
          XM_005269572.3 XP_005269629.1 6 922
          XM_006717642.2 XP_006717705.1 9 1348
          XM_011539349.2 XP_011537651.1 5 1130
          XM_011539351.1 XP_011537653.1 10 1449
          XM_011539353.1 XP_011537655.1 9 1320
          XM_011539354.1 XP_011537656.1 8 1244
          XM_011539355.1 XP_011537657.1 7 1322
          XM_011539357.1 XP_011537659.1 7 1060
          XM_011539358.1 XP_011537660.1 6 1138
          XM_017015756.1 XP_016871245.1 8 1090
          XM_017015757.1 XP_016871246.1 7 2816
          GenBank mRNA AF016507.1 - 7 953
          AF222711.1 - 5 2370
          AK290390.1 - 7 897
          AL833398.1 - 7 1008
          BC002486.2 - 7 1009
          BC037900.2 - 5 1017 5276086
          BC047018.1 - 7 1013
          BC052276.1 - 7 990
          BC072020.1 - 7 970
          BT007012.1 - 5 608
          DB200827.1 - 5 581

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_053335, AF222712.
          This siRNA also targets the following transcripts in Mouse: NM_001170744, NM_009980, AK147871, BC017520, BC013333, and others.

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          negative regulation of cell proliferation
          viral genome replication
          positive regulation of chromatin binding
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of retinoic acid receptor signaling pathway
          white fat cell differentiation
          oxidation-reduction process
          chromatin binding
          transcription coactivator activity
          transcription corepressor activity
          protein binding
          oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          protein homodimerization activity
          retinoic acid receptor binding
          NAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0