Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152513.3 | NP_689726.3 | 12 | 1410 | ||
XM_011529935.1 | XP_011528237.1 | 12 | 1419 | |||
XM_011529936.1 | XP_011528238.1 | 12 | 1419 | |||
XM_011529937.1 | XP_011528239.1 | 12 | 1419 | |||
XM_011529938.1 | XP_011528240.1 | 11 | 1302 | |||
XM_011529939.1 | XP_011528241.1 | 12 | 1419 | |||
XM_011529940.1 | XP_011528242.1 | 12 | 1419 | |||
XM_011529941.1 | XP_011528243.1 | 12 | 1419 | |||
XM_011529942.2 | XP_011528244.1 | 12 | 1610 | |||
XM_011529943.1 | XP_011528245.1 | 12 | 1419 | |||
XM_011529944.1 | XP_011528246.1 | 12 | 1419 | |||
XM_011529945.2 | XP_011528247.1 | 12 | 1421 | |||
XM_011529948.2 | XP_011528250.1 | 8 | 791 | |||
XM_011529949.1 | XP_011528251.1 | 7 | 728 | |||
XM_011529950.1 | XP_011528252.1 | 12 | 1419 | |||
XM_011529952.1 | XP_011528254.1 | 7 | 865 | |||
XM_011529954.1 | XP_011528256.1 | 2 | 143 | |||
XM_011529955.1 | XP_011528257.1 | 12 | 1419 | |||
XM_011529956.2 | XP_011528258.1 | 12 | 1419 | |||
XM_017028633.1 | XP_016884122.1 | 12 | 1419 | |||
GenBank mRNA | BC025400.1 | - | 12 | 1426 | ||
BC070111.1 | - | 7 | 867 |