Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKAR SIRNA Assays ›
Gene Symbol:
ANKAR
Gene Name:
ankyrin and armadillo repeat containing
Gene Aliases:
-
Chromosome Location:
Chr.2: 189674292 - 189763059 on Build GRCh38
UniGene ID:
Hs.60772
Species:
Human
siRNA ID
HSS175638
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKAR
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_144708.3
NP_653309.3
19
3716
XM_011510673.2
XP_011508975.1
19
4235
XM_011510674.1
XP_011508976.1
19
4202
XM_011510675.2
XP_011508977.1
18
4130
XM_011510676.2
XP_011508978.1
19
4235
XM_011510677.2
XP_011508979.1
19
4235
XM_011510679.2
XP_011508981.1
17
3806
XM_011510680.1
XP_011508982.1
19
4235
XM_011510681.1
XP_011508983.1
19
4235
XM_011510682.2
XP_011508984.1
19
4235
XM_011510685.2
XP_011508987.1
19
3791
XM_011510687.2
XP_011508989.1
18
3599
XM_011510691.2
XP_011508993.1
15
2421
XM_011510692.2
XP_011508994.1
17
3435
XM_011510693.2
XP_011508995.1
16
3331
XM_011510694.2
XP_011508996.1
15
3150
XM_017003413.1
XP_016858902.1
19
4235
XM_017003415.1
XP_016858904.1
14
2930
XM_017003416.1
XP_016858905.1
16
3342
XM_017003418.1
XP_016858907.1
14
3045
XM_017003419.1
XP_016858908.1
14
3012
XM_017003420.1
XP_016858909.1
14
2948
GenBank mRNA
AJ549812.1
-
18
3415
AJ605761.1
-
7
884
AK058144.1
-
7
1068
AK126073.1
-
17
3486
BC037215.1
-
13
2705
BC044907.1
-
15
3209
4836761
BC047413.1
-
22
3280
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-tvqvm:80/10.239.167.222:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0