Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142300.1 | NP_001135772.1 | 5 | 823 | ||
NM_152523.2 | NP_689736.1 | 8 | 724 | |||
XM_005246343.1 | XP_005246400.1 | 7 | 742 | |||
GenBank mRNA | AK095479.1 | - | 2 | 163 | ||
AK097751.1 | - | 8 | 724 | |||
BC067253.1 | - | 5 | 610 | 6500304 |