Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CTSL SIRNA Assays ›
          Gene Symbol:
          CTSL
          Gene Name:
          cathepsin L
          Gene Aliases:
          CATL, CTSL1, MEP
          Chromosome Location:
          Chr.9: 87725437 - 87731469 on Build GRCh38
          UniGene ID:
          Hs.731507
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS175681
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CTSL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001257971.1 NP_001244900.1 2 334
          NM_001257972.1 NP_001244901.1 2 275
          NM_001257973.1 NP_001244902.1 2 279
          NM_001912.4 NP_001903.1 2 424
          NM_145918.2 NP_666023.1 2 279
          XM_005251716.3 XP_005251773.1 3 1154
          XM_011518263.1 XP_011516565.1 2 964
          XM_017014293.1 XP_016869782.1 2 901
          GenBank mRNA AF217997.1 - 2 204
          AF304301.1 - 2 218
          AF467444.1 - 2 273
          AK055599.1 - 2 964
          AK075100.1 - 2 230
          AL832167.1 - 2 222
          BC012612.1 - 2 272 4295635
          BC142983.1 - 2 219 8860422
          BI824449.1 - 2 273
          BI829385.1 - 2 378
          BX537395.1 - 2 215
          BX647101.1 - 2 219
          BX647102.1 - 2 218
          BX647413.1 - 2 219
          BX647434.1 - 2 277
          BX647435.1 - 2 274
          BX648848.1 - 2 229
          BX648849.1 - 2 219
          CR457053.1 - 1 74
          X12451.1 - 2 362

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          toll-like receptor signaling pathway
          adaptive immune response
          proteolysis
          antigen processing and presentation
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          extracellular matrix disassembly
          collagen catabolic process
          proteolysis involved in cellular protein catabolic process
          macrophage apoptotic process
          cellular response to thyroid hormone stimulus
          fibronectin binding
          cysteine-type endopeptidase activity
          serine-type endopeptidase activity
          protein binding
          collagen binding
          cysteine-type peptidase activity
          histone binding
          proteoglycan binding
          serpin family protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0