Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CYP1A1 SIRNA Assays ›
          Gene Symbol:
          CYP1A1
          Gene Name:
          cytochrome P450 family 1 subfamily A member 1
          Gene Aliases:
          AHH, AHRR, CP11, CYP1, CYPIA1, P1-450, P450-C, P450DX
          Chromosome Location:
          Chr.15: 74719542 - 74725610 on Build GRCh38
          UniGene ID:
          Hs.72912
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS175787
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CYP1A1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000499.4 NP_000490.1 3 1107
          NM_001319216.1 NP_001306145.1 3 1110
          NM_001319217.1 NP_001306146.1 3 1110
          XM_017021953.1 XP_016877442.1 4 1026
          GenBank mRNA AK223108.1 - 3 1028
          AK223113.1 - 3 1025
          AK310810.1 - 3 1154
          AK313880.1 - 3 1025
          AM233517.1 - 3 399
          AM233518.1 - 3 399
          AM233519.1 - 4 795
          AM233520.1 - 4 1145
          AM236046.1 - 4 911
          AM236047.1 - 3 206
          AY310359.1 - 3 827
          AY871801.1 - 3 994
          BC023019.1 - 3 992 5123393
          K03191.1 - 3 997
          M12079.1 - 1 29

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          porphyrin-containing compound metabolic process
          aging
          cell proliferation
          amine metabolic process
          response to wounding
          response to virus
          response to nematode
          response to herbicide
          ethylene metabolic process
          embryo development ending in birth or egg hatching
          coumarin metabolic process
          flavonoid metabolic process
          response to iron(III) ion
          insecticide metabolic process
          drug metabolic process
          dibenzo-p-dioxin catabolic process
          epoxygenase P450 pathway
          response to food
          response to lipopolysaccharide
          response to vitamin A
          response to immobilization stress
          vitamin D metabolic process
          response to drug
          9-cis-retinoic acid biosynthetic process
          camera-type eye development
          response to antibiotic
          response to arsenic-containing substance
          digestive tract development
          hydrogen peroxide biosynthetic process
          response to hyperoxia
          oxidation-reduction process
          maternal process involved in parturition
          hepatocyte differentiation
          demethylation
          cellular response to organic cyclic compound
          omega-hydroxylase P450 pathway
          positive regulation of G1/S transition of mitotic cell cycle
          monooxygenase activity
          iron ion binding
          protein binding
          steroid hydroxylase activity
          oxidoreductase activity
          oxidoreductase activity, acting on diphenols and related substances as donors
          flavonoid 3'-monooxygenase activity
          oxygen binding
          enzyme binding
          heme binding
          demethylase activity
          aromatase activity
          vitamin D 24-hydroxylase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-26xzd:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0