Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DAXX SIRNA Assays ›
          Gene Symbol:
          DAXX
          Gene Name:
          death domain associated protein
          Gene Aliases:
          BING2, DAP6, EAP1
          Chromosome Location:
          Chr.6: 33318558 - 33323016 on Build GRCh38
          UniGene ID:
          Hs.336916
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS175937
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DAXX
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001141969.1 NP_001135441.1 2 365
          NM_001141970.1 NP_001135442.1 2 296
          NM_001350.4 NP_001341.1 2 362
          GenBank mRNA AB209493.1 - 1 380
          AF015956.2 - 1 212
          AF039136.1 - 2 305
          AF050179.1 - 2 273
          AF097742.1 - 1 165
          AK223404.1 - 2 363
          AK292187.1 - 2 363
          AK303767.1 - 2 239
          AK309612.1 - 2 264
          AK313034.1 - 2 305
          BC109073.1 - 2 248 40034214
          BC109074.1 - 2 248 40034219
          CR457085.1 - 1 158
          DC345308.1 - 2 362
          DC359186.1 - 2 365
          HQ436528.1 - 1 158
          HQ436529.1 - 1 158

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          positive regulation of protein phosphorylation
          nucleosome assembly
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          apoptotic process
          activation of JUN kinase activity
          extrinsic apoptotic signaling pathway via death domain receptors
          response to metal ion
          negative regulation of myotube differentiation
          viral process
          covalent chromatin modification
          androgen receptor signaling pathway
          PML body organization
          regulation of protein ubiquitination
          positive regulation of histone phosphorylation
          positive regulation of apoptotic process
          positive regulation of protein kinase activity
          negative regulation of transcription, DNA-templated
          cellular response to tumor necrosis factor
          cellular response to anoxia
          positive regulation of neuron death
          p53 binding
          transcription corepressor activity
          receptor signaling protein activity
          protein binding
          transcription factor binding
          enzyme binding
          protein kinase binding
          protein kinase activator activity
          heat shock protein binding
          ubiquitin protein ligase binding
          histone binding
          protein N-terminus binding
          androgen receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qlhjl:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0