Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DLG4 SIRNA Assays ›
          Gene Symbol:
          DLG4
          Gene Name:
          discs large MAGUK scaffold protein 4
          Gene Aliases:
          PSD95, SAP-90, SAP90
          Chromosome Location:
          Chr.17: 7189890 - 7220050 on Build GRCh38
          UniGene ID:
          Hs.463928
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS176193
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DLG4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001128827.2 NP_001122299.1 9 1471
          NM_001321074.1 NP_001308003.1 11 2319
          NM_001321075.1 NP_001308004.1 9 1480
          NM_001321076.1 NP_001308005.1 8 1125
          NM_001321077.1 NP_001308006.1 8 1116
          NM_001365.4 NP_001356.1 11 2328
          NR_135527.1 11 2328
          XM_005256491.1 XP_005256548.1 9 1570
          XM_011523698.1 XP_011522000.1 11 2216
          XM_011523699.2 XP_011522001.1 12 1458
          XM_011523702.1 XP_011522004.1 8 995
          XM_017024288.1 XP_016879777.1 8 1216
          XM_017024289.1 XP_016879778.1 8 1207
          XM_017024290.1 XP_016879779.1 8 1685
          GenBank mRNA AK293835.1 - 9 1477
          AK296357.1 - 11 2016
          AK297346.1 - 8 1073
          AK299801.1 - 8 1087
          AK308533.1 - 8 1230
          AK316096.1 - 9 1033
          BC040533.1 - 3 473
          BC136553.1 - 11 1852 9020543
          U68138.1 - 7 887
          U83192.1 - 11 1986

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_007864, NM_001109752, BC014807, D50621, BQ959940.

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          negative regulation of receptor internalization
          protein complex assembly
          signal transduction
          positive regulation of cytosolic calcium ion concentration
          chemical synaptic transmission
          nervous system development
          learning
          synaptic vesicle maturation
          ion transmembrane transport
          social behavior
          protein localization to synapse
          locomotory exploration behavior
          positive regulation of GTPase activity
          establishment of protein localization
          establishment or maintenance of epithelial cell apical/basal polarity
          GMP metabolic process
          GDP metabolic process
          regulation of long-term neuronal synaptic plasticity
          positive regulation of synaptic transmission
          neuromuscular process controlling balance
          dendritic spine morphogenesis
          vocalization behavior
          AMPA glutamate receptor clustering
          receptor localization to synapse
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          positive regulation of excitatory postsynaptic potential
          regulation of grooming behavior
          guanylate kinase activity
          Ras guanyl-nucleotide exchange factor activity
          protein binding
          protein C-terminus binding
          ligand-gated ion channel activity
          kinase binding
          protein phosphatase binding
          PDZ domain binding
          beta-1 adrenergic receptor binding
          D1 dopamine receptor binding
          P2Y1 nucleotide receptor binding
          protein complex binding
          acetylcholine receptor binding
          ionotropic glutamate receptor binding
          neuroligin family protein binding
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-hltnz:80/100.66.72.22:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0