Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DTNB SIRNA Assays ›
Gene Symbol:
DTNB
Gene Name:
dystrobrevin beta
Gene Aliases:
-
Chromosome Location:
Chr.2: 25377220 - 25673647 on Build GRCh38
UniGene ID:
Hs.307720
Species:
Human
siRNA ID
HSS176263
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DTNB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256303.1
NP_001243232.1
6
809
NM_001256304.1
NP_001243233.1
5
546
NM_001256308.2
NP_001243237.1
5
680
NM_001320933.1
NP_001307862.1
6
809
NM_001320934.1
NP_001307863.1
6
809
NM_001320936.1
NP_001307865.1
6
809
NM_001320937.1
NP_001307866.1
6
809
NM_021907.4
NP_068707.1
6
809
NM_033147.3
NP_149159.2
6
809
NM_033148.3
NP_149160.1
6
809
NM_183360.2
NP_899204.1
6
809
NM_183361.2
NP_899205.1
6
809
XM_005264182.1
XP_005264239.1
6
726
XM_011532671.1
XP_011530973.1
6
726
XM_011532672.2
XP_011530974.1
6
726
XM_011532673.1
XP_011530975.1
6
726
XM_011532674.1
XP_011530976.1
6
726
XM_011532675.1
XP_011530977.1
6
726
XM_011532676.1
XP_011530978.1
6
726
XM_011532677.2
XP_011530979.1
5
1182
XM_011532679.1
XP_011530981.1
6
726
XM_011532680.2
XP_011530982.1
5
647
XM_011532681.2
XP_011530983.1
5
640
XM_017003529.1
XP_016859018.1
6
726
XM_017003530.1
XP_016859019.1
6
726
XM_017003531.1
XP_016859020.1
6
726
XM_017003532.1
XP_016859021.1
6
726
XM_017003533.1
XP_016859022.1
6
726
XM_017003534.1
XP_016859023.1
6
726
XM_017003535.1
XP_016859024.1
6
726
XM_017003536.1
XP_016859025.1
6
726
XM_017003537.1
XP_016859026.1
2
1079
XM_017003538.1
XP_016859027.1
5
572
XM_017003539.1
XP_016859028.1
5
583
XM_017003540.1
XP_016859029.1
5
640
XM_017003541.1
XP_016859030.1
5
624
GenBank mRNA
AF022728.1
-
6
707
AI608704.1
-
4
545
AK094494.1
-
6
672
AK294357.1
-
5
490
AK300019.1
-
6
686
AK301386.1
-
5
682
AL555818.3
-
6
723
BC016655.1
-
6
702
3868207
BC049366.1
-
6
800
4817552
BT009805.1
-
5
546
BU846095.1
-
4
487
DQ160290.1
-
5
546
Y12712.1
-
6
729
Y15718.1
-
5
546
Y15722.1
-
5
546
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Process(es)
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7dbcb7bc98-zbtnl:80/10.239.166.103:80.
git-commit: 8789124447060be43cea05543d36199cab59d3d8
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.5.0-2024.03.10-1.0