Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RILPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145058.2 | NP_659495.1 | 2 | 686 | ||
XM_011538012.2 | XP_011536314.1 | 2 | 630 | |||
XM_017018959.1 | XP_016874448.1 | 2 | 630 | |||
GenBank mRNA | AB085763.1 | - | 2 | 449 | ||
AK056934.1 | - | 2 | 670 | |||
BC013042.2 | - | 2 | 459 | 3544761 |