Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000534.1 | XP_016856023.1 | 16 | 3018 | ||
XM_017000535.1 | XP_016856024.1 | 16 | 2978 | |||
XM_017000536.1 | XP_016856025.1 | 15 | 2826 | |||
GenBank mRNA | AK296788.1 | - | 15 | 2071 | ||
BC008655.1 | - | 4 | 580 | |||
BC037166.2 | - | 16 | 2866 | 5090654 | ||
M59371.1 | - | 16 | 2861 |