Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: THAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152658.2 | NP_689871.1 | 3 | 1157 | ||
XM_005258623.2 | XP_005258680.1 | 3 | 966 | |||
XM_011526604.2 | XP_011524906.1 | 3 | 1166 | |||
GenBank mRNA | AK057453.1 | - | 4 | 1013 | ||
AK093048.1 | - | 3 | 1158 | |||
AK296640.1 | - | 3 | 651 | |||
AK297630.1 | - | 2 | 467 | |||
BC046207.1 | - | 3 | 706 | 6176484 | ||
BC072416.1 | - | 3 | 797 | 6169558 |