Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305242.1 | NP_001292171.1 | 2 | 279 | ||
NM_144988.3 | NP_659425.1 | 2 | 279 | |||
XM_005270582.3 | XP_005270639.1 | 2 | 301 | |||
XM_011540897.2 | XP_011539199.1 | 2 | 301 | |||
GenBank mRNA | AK289395.1 | - | 2 | 279 | ||
AW242346.1 | - | 2 | 193 | |||
BC011706.2 | - | 2 | 255 | 3689162 |