Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173551.4 | NP_775822.3 | 5 | 1209 | ||
XM_005251793.4 | XP_005251850.1 | 5 | 1196 | |||
XM_005251794.4 | XP_005251851.1 | 5 | 1196 | |||
XM_006716998.3 | XP_006717061.1 | 5 | 1196 | |||
XM_006716999.2 | XP_006717062.1 | 5 | 2281 | |||
XM_011518357.2 | XP_011516659.1 | 5 | 3278 | |||
XM_017014445.1 | XP_016869934.1 | 5 | 1196 | |||
XM_017014446.1 | XP_016869935.1 | 4 | 3157 | |||
XM_017014447.1 | XP_016869936.1 | 1 | 31 | |||
GenBank mRNA | AK126614.1 | - | 3 | 263 | ||
AK127702.1 | - | 4 | 796 | |||
BC064367.1 | - | 2 | 196 | 5562895 | ||
CR749467.1 | - | 5 | 880 | |||
CR749472.1 | - | 3 | 560 |