Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EPB41L1 SIRNA Assays ›
Gene Symbol:
EPB41L1
Gene Name:
erythrocyte membrane protein band 4.1 like 1
Gene Aliases:
4.1N, MRD11
Chromosome Location:
Chr.20: 36064891 - 36232799 on Build GRCh38
UniGene ID:
Hs.437422
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS176579
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EPB41L1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258329.1
NP_001245258.1
9, 10
1103
NM_001258330.1
NP_001245259.1
9, 10
965
NM_001258331.1
NP_001245260.1
8, 9
865
NM_012156.2
NP_036288.2
8, 9
1049
NM_177996.2
NP_818932.1
8, 9
860
XM_011528667.1
XP_011526969.1
13, 14
1995
XM_011528671.1
XP_011526973.1
9, 10
1082
XM_011528673.1
XP_011526975.1
8, 9
1042
XM_011528677.1
XP_011526979.1
8, 9
856
XM_011528681.1
XP_011526983.1
7, 8
892
XM_011528682.1
XP_011526984.1
7, 8
892
XM_011528683.1
XP_011526985.1
7, 8
892
XM_011528685.1
XP_011526987.1
7, 8
892
XM_017027711.1
XP_016883200.1
7, 8
805
XM_017027712.1
XP_016883201.1
8, 9
943
XM_017027713.1
XP_016883202.1
8, 9
922
XM_017027714.1
XP_016883203.1
8, 9
919
XM_017027715.1
XP_016883204.1
7, 8
892
XM_017027716.1
XP_016883205.1
9, 10
1082
XM_017027717.1
XP_016883206.1
8, 9
1040
XM_017027718.1
XP_016883207.1
8, 9
919
XM_017027719.1
XP_016883208.1
8, 9
894
XM_017027720.1
XP_016883209.1
10, 11
1153
GenBank mRNA
AB002336.1
-
8, 9
1039
AK096848.1
-
8, 9
912
AK126875.1
-
9, 10
963
AK299817.1
-
8, 9
996
AY049789.1
-
7, 8
887
BC013885.2
-
8, 9
865
3688606
BC040259.1
-
9, 10
1081
5195986
CR936825.1
-
8, 9
887
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cortical actin cytoskeleton organization
cell-cell adhesion
actin binding
structural molecule activity
protein binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline