Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other F2R SIRNA Assays ›
          Gene Symbol:
          F2R
          Gene Name:
          coagulation factor II thrombin receptor
          Gene Aliases:
          CF2R, HTR, PAR-1, PAR1, TR
          Chromosome Location:
          Chr.5: 76716043 - 76735780 on Build GRCh38
          UniGene ID:
          Hs.482562
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS176656
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: F2R
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001311313.1 NP_001298242.1 3 732
          NM_001992.4 NP_001983.2 2 610
          GenBank mRNA AK308169.1 - 2 610
          AK314202.1 - 2 533
          BC002464.2 - 2 525 3343051
          BC051909.1 - 2 531 6172703
          BT007279.1 - 2 345
          M62424.1 - 2 569

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPKK activity
          connective tissue replacement involved in inflammatory response wound healing
          negative regulation of glomerular filtration
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          inflammatory response
          G-protein coupled receptor signaling pathway
          phospholipase C-activating G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          protein kinase C-activating G-protein coupled receptor signaling pathway
          tyrosine phosphorylation of STAT protein
          STAT protein import into nucleus
          establishment of synaptic specificity at neuromuscular junction
          blood coagulation
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          response to wounding
          anatomical structure morphogenesis
          positive regulation of phosphatidylinositol 3-kinase signaling
          platelet activation
          regulation of blood coagulation
          positive regulation of blood coagulation
          positive regulation of cell migration
          response to lipopolysaccharide
          regulation of interleukin-1 beta production
          positive regulation of collagen biosynthetic process
          positive regulation of Rho protein signal transduction
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of MAPK cascade
          negative regulation of neuron apoptotic process
          positive regulation of transcription, DNA-templated
          positive regulation of vasoconstriction
          positive regulation of smooth muscle contraction
          positive regulation of JAK-STAT cascade
          homeostasis of number of cells within a tissue
          release of sequestered calcium ion into cytosol
          positive regulation of release of sequestered calcium ion into cytosol
          positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
          positive regulation of calcium ion transport
          regulation of sensory perception of pain
          platelet dense granule organization
          positive regulation of ERK1 and ERK2 cascade
          thrombin receptor signaling pathway
          negative regulation of renin secretion into blood stream
          positive regulation of interleukin-8 secretion
          positive regulation of interleukin-6 secretion
          G-protein alpha-subunit binding
          G-protein coupled receptor activity
          receptor binding
          protein binding
          thrombin receptor activity
          G-protein beta-subunit binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-s97cd:80/100.66.77.220:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0