Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291992.1 | NP_001278921.1 | 3 | 295 | ||
NM_174927.2 | NP_777587.1 | 3 | 295 | |||
GenBank mRNA | AF521713.1 | - | 3 | 231 | ||
AK098717.1 | - | 3 | 254 | |||
BC058039.1 | - | 3 | 262 | 6619434 | ||
BX096445.1 | - | 3 | 295 |