Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001971.2 | NP_001001971.1 | 10 | 1255 | ||
NM_001143773.1 | NP_001137245.1 | 13 | 1720 | |||
NM_001166698.1 | NP_001160170.1 | 13 | 1631 | |||
NM_198215.3 | NP_937858.2 | 12 | 1549 | |||
XM_005269618.4 | XP_005269675.2 | 12 | 1546 | |||
XM_005269619.4 | XP_005269676.2 | 10 | 1252 | |||
XM_006717702.3 | XP_006717765.1 | 14 | 1550 | |||
XM_006717703.3 | XP_006717766.1 | 14 | 1732 | |||
XM_011539490.2 | XP_011537792.1 | 13 | 1605 | |||
XM_011539491.2 | XP_011537793.1 | 14 | 1651 | |||
XM_017015885.1 | XP_016871374.1 | 13 | 1463 | |||
XM_017015886.1 | XP_016871375.1 | 13 | 1717 | |||
XM_017015887.1 | XP_016871376.1 | 13 | 1467 | |||
XM_017015888.1 | XP_016871377.1 | 14 | 1729 | |||
XM_017015889.1 | XP_016871378.1 | 14 | 1547 | |||
XM_017015890.1 | XP_016871379.1 | 14 | 1786 | |||
XM_017015891.1 | XP_016871380.1 | 11 | 1426 | |||
XM_017015892.1 | XP_016871381.1 | 11 | 1423 | |||
XM_017015893.1 | XP_016871382.1 | 13 | 1527 | |||
GenBank mRNA | AK289796.1 | - | 12 | 1475 | ||
AK294805.1 | - | 12 | 1464 | |||
AK299924.1 | - | 13 | 1720 | |||
AK316542.1 | - | 13 | 1627 | |||
BC036453.1 | - | 12 | 1507 | 5270033 | ||
BC064431.1 | - | 10 | 1255 | 5764753 |