Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FHL1 SIRNA Assays ›
Gene Symbol:
FHL1
Gene Name:
four and a half LIM domains 1
Gene Aliases:
FHL-1, FHL1A, FHL1B, FLH1A, KYOT, RBMX1A, RBMX1B, SLIM, SLIM-1, SLIM1, SLIMMER, XMPMA
Chromosome Location:
Chr.X: 136146702 - 136211359 on Build GRCh38
UniGene ID:
Hs.435369
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
HSS176931
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FHL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001159699.1
NP_001153171.1
2, 3
421
NM_001159700.1
NP_001153172.1
3, 4
416
NM_001159701.1
NP_001153173.1
2, 3
292
NM_001159702.2
NP_001153174.1
3, 4
485
NM_001159703.1
NP_001153175.1
3, 4
485
NM_001159704.1
NP_001153176.1
2, 3
867
NM_001167819.1
NP_001161291.1
3, 4
353
NM_001449.4
NP_001440.2
3, 4
485
NR_027621.1
3, 4
567
XM_006724743.2
XP_006724806.1
2, 3
289
XM_006724744.2
XP_006724807.1
3, 4
367
XM_006724745.3
XP_006724808.1
3, 4
305
XM_006724746.2
XP_006724809.1
2, 3
240
XM_006724747.2
XP_006724810.1
2, 3
240
XM_011531316.1
XP_011529618.1
2, 3
289
XM_017029357.1
XP_016884846.1
3, 4
367
GenBank mRNA
AF063002.1
-
2, 3
240
AF098518.1
-
2, 3
209
AF220153.1
-
1, 2
156
AK122708.1
-
2, 3
868
AK223299.1
-
1, 2
190
AK289411.1
-
3, 4
567
AK299381.1
-
2, 3
267
AK299450.1
-
2, 3
264
AK300966.1
-
4, 5
292
AK301642.1
-
2, 3
289
AK304680.1
-
3, 4
344
BC010998.2
-
3, 4
364
4039973
BC088369.1
-
2, 3
254
30529295
BE965488.2
-
3, 4
293
CR456974.1
-
1, 2
156
DA497183.1
-
3, 4
485
DA917361.1
-
1, 2
51
DA982372.1
-
3, 4
354
DC363191.1
-
3, 4
416
DC373304.1
-
2, 3
293
DC373386.1
-
2, 3
422
U29538.1
-
1, 2
177
U60115.1
-
2, 3
239
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of membrane depolarization
muscle organ development
organ morphogenesis
negative regulation of G2/M transition of mitotic cell cycle
cell differentiation
negative regulation of cell growth
positive regulation of potassium ion transport
regulation of potassium ion transmembrane transporter activity
negative regulation of G1/S transition of mitotic cell cycle
molecular_function
protein binding
zinc ion binding
ion channel binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0