Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FHL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004468.4 | NP_004459.2 | 2 | 305 | ||
GenBank mRNA | AF133732.1 | - | 1 | 126 | ||
AK290641.1 | - | 2 | 305 | |||
AK309179.1 | - | 2 | 222 | |||
BC001351.2 | - | 2 | 295 | 2964682 | ||
BC011697.2 | - | 2 | 295 | 3532981 | ||
BT007052.1 | - | 1 | 126 | |||
BX384389.2 | - | 2 | 361 | |||
CR457425.1 | - | 1 | 126 | |||
U60116.2 | - | 1 | 165 |