Search
Search

NID2| Gene Symbol: NID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007361.3 | NP_031387.3 | 13 | 2948 | ||
| XM_005267405.4 | XP_005267462.1 | 12 | 3032 | |||
| XM_005267406.4 | XP_005267463.1 | 11 | 2888 | |||
| XM_005267407.4 | XP_005267464.1 | 12 | 2807 | |||
| GenBank mRNA | AJ223500.1 | - | 13 | 2714 | ||
| AK291652.1 | - | 13 | 2776 | |||
| AK298542.1 | - | 4 | 519 | |||
| AK300462.1 | - | 13 | 2442 | |||
| BC035608.1 | - | 9 | 1550 | 5205293 | ||
| BX648241.1 | - | 14 | 2721 | |||
| D86425.1 | - | 13 | 2717 | |||