Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FKBP1B SIRNA Assays ›
          Gene Symbol:
          FKBP1B
          Gene Name:
          FK506 binding protein 1B
          Gene Aliases:
          FKBP12.6, FKBP1L, OTK4, PKBP1L, PPIase
          Chromosome Location:
          Chr.2: 24033205 - 24067743 on Build GRCh38
          UniGene ID:
          Hs.709461
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS176952
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FKBP1B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001322963.1 NP_001309892.1 4 343
          NM_001322964.1 NP_001309893.1 3 162
          NM_004116.4 NP_004107.1 3 275
          NM_054033.3 NP_473374.1 3 275
          NR_136536.1 5 432
          NR_136538.1 4 364
          NR_136539.1 3 162
          XM_011532699.2 XP_011531001.1 3 224
          XM_017003593.1 XP_016859082.1 5 378
          XM_017003594.1 XP_016859083.1 3 142
          XM_017003595.1 XP_016859084.1 2 123
          XM_017003596.1 XP_016859085.1 4 369
          GenBank mRNA AF322070.1 - 3 167
          BC002614.1 - 3 218 3161896
          BF216711.1 - 3 130
          BF969580.1 - 3 262
          BI546013.1 - 3 240
          BM802489.1 - 3 275
          BU147063.1 - 3 113
          BU508033.1 - 3 239
          D38037.1 - 3 198
          L37086.1 - 3 192
          S69800.1 - 3 198
          S69815.1 - 3 198

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_016863, BC061121, BE861051, AF060872, AB285017, and others.

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          'de novo' protein folding
          smooth muscle contraction
          response to glucose
          negative regulation of heart rate
          regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
          release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          neuronal action potential propagation
          protein maturation by protein folding
          insulin secretion
          negative regulation of protein phosphatase type 2B activity
          response to vitamin E
          ion transmembrane transport
          calcium-mediated signaling using intracellular calcium source
          protein refolding
          T cell proliferation
          response to hydrogen peroxide
          positive regulation of axon regeneration
          negative regulation of release of sequestered calcium ion into cytosol
          positive regulation of sequestering of calcium ion
          regulation of cytosolic calcium ion concentration
          response to redox state
          regulation of ryanodine-sensitive calcium-release channel activity
          negative regulation of ryanodine-sensitive calcium-release channel activity
          chaperone-mediated protein folding
          negative regulation of insulin secretion involved in cellular response to glucose stimulus
          cell communication by electrical coupling involved in cardiac conduction
          regulation of cardiac conduction
          peptidyl-prolyl cis-trans isomerase activity
          receptor binding
          ryanodine-sensitive calcium-release channel activity
          protein binding
          FK506 binding
          calcium channel inhibitor activity
          cyclic nucleotide binding
          ion channel binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0