Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014920.3 | NP_055735.1 | 12 | 1980 | ||
NM_016513.4 | NP_057597.2 | 13 | 2091 | |||
XM_011514419.1 | XP_011512721.1 | 12 | 2001 | |||
XM_011514420.2 | XP_011512722.1 | 13 | 2143 | |||
XM_011514421.1 | XP_011512723.1 | 13 | 2082 | |||
XM_017010485.1 | XP_016865974.1 | 13 | 2375 | |||
XM_017010486.1 | XP_016865975.1 | 14 | 2254 | |||
XM_017010487.1 | XP_016865976.1 | 12 | 1920 | |||
XM_017010488.1 | XP_016865977.1 | 13 | 2781 | |||
XM_017010489.1 | XP_016865978.1 | 14 | 2296 | |||
XM_017010490.1 | XP_016865979.1 | 13 | 2166 | |||
XM_017010491.1 | XP_016865980.1 | 13 | 2127 | |||
XM_017010492.1 | XP_016865981.1 | 12 | 2171 | |||
GenBank mRNA | AB023153.1 | - | 12 | 1867 | ||
AF152469.1 | - | 12 | 1948 | |||
AF225919.1 | - | 12 | 1810 | |||
AK074892.1 | - | 13 | 2061 | |||
BC136420.1 | - | 11 | 1641 | 9020408 | ||
BC136421.1 | - | 11 | 1641 | 9020409 | ||
BC152464.1 | - | 12 | 1867 | 8862600 |