Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142550.1 | NP_001136022.1 | 14, 15 | 3028 | ||
NM_001142551.1 | NP_001136023.1 | 14, 15 | 3004 | |||
NM_014969.5 | NP_055784.3 | 14, 15 | 3007 | |||
XM_006710460.3 | XP_006710523.1 | 14, 15 | 3005 | |||
XM_011541027.2 | XP_011539329.1 | 14, 15 | 3023 | |||
XM_011541028.2 | XP_011539330.1 | 14, 15 | 3002 | |||
XM_011541029.1 | XP_011539331.1 | 14, 15 | 3026 | |||
XM_011541030.1 | XP_011539332.1 | 14, 15 | 2674 | |||
XM_011541031.1 | XP_011539333.1 | 12, 13 | 2501 | |||
XM_017000696.1 | XP_016856185.1 | 14, 15 | 3017 | |||
XM_017000697.1 | XP_016856186.1 | 14, 15 | 2665 | |||
GenBank mRNA | AB020700.1 | - | 14, 15 | 2851 | ||
AK225781.1 | - | 14, 15 | 2788 | |||
AK289789.1 | - | 14, 15 | 2881 | |||
AK294998.1 | - | 11, 12 | 2394 | |||
BC034964.2 | - | 14, 15 | 2892 | 4823902 | ||
BC039254.1 | - | 13, 14 | 2800 | 5287954 |