Search
Search

KIAA0922| Gene Symbol: KIAA0922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131007.1 | NP_001124479.1 | 25 | 3092 | ||
| NM_015196.3 | NP_056011.3 | 25 | 3089 | |||
| XM_011531780.1 | XP_011530082.1 | 25 | 3142 | |||
| XM_011531781.1 | XP_011530083.1 | 25 | 3139 | |||
| XM_011531783.1 | XP_011530085.1 | 23 | 2890 | |||
| XM_017007924.1 | XP_016863413.1 | 24 | 3064 | |||
| XM_017007925.1 | XP_016863414.1 | 25 | 3092 | |||
| XM_017007926.1 | XP_016863415.1 | 25 | 3089 | |||
| XM_017007927.1 | XP_016863416.1 | 8 | 1197 | |||
| GenBank mRNA | AB023139.1 | - | 3 | 580 | ||
| AK001454.1 | - | 1 | 115 | |||
| AK092270.1 | - | 25 | 3089 | |||
| AK096538.1 | - | 5 | 909 | |||
| AL136932.1 | - | 14 | 2049 | |||
| BC044932.1 | - | 18 | 2371 | |||
| BC131505.1 | - | 24 | 2861 | 40082592 | ||