Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204204.2 | NP_001191133.1 | 4 | 366 | ||
NM_001291932.1 | NP_001278861.1 | 5 | 539 | |||
NM_012429.4 | NP_036561.1 | 6 | 615 | |||
NM_033382.2 | NP_203740.1 | 6 | 615 | |||
GenBank mRNA | AB033012.2 | - | 6 | 514 | ||
AK056163.1 | - | 1 | 91 | |||
AK091402.1 | - | 4 | 402 | |||
AK223587.1 | - | 6 | 575 | |||
AK296540.1 | - | 5 | 429 | |||
AK303751.1 | - | 4 | 262 | |||
AK314390.1 | - | 6 | 548 | |||
AL096881.1 | - | 6 | 612 | |||
AY927438.1 | - | 5 | 344 | |||
AY927439.1 | - | 5 | 344 | |||
BC058915.1 | - | 6 | 519 | 5178251 | ||
CR456571.1 | - | 6 | 512 | |||
DC375707.1 | - | 5 | 540 |