Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RBFOX2 SIRNA Assays ›
          Gene Symbol:
          RBFOX2
          Gene Name:
          RNA binding protein, fox-1 homolog 2
          Gene Aliases:
          FOX2, Fox-2, HNRBP2, HRNBP2, RBM9, RTA, dJ106I20.3, fxh
          Chromosome Location:
          Chr.22: 35738736 - 36028892 on Build GRCh38
          UniGene ID:
          Hs.282998
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS177482
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RBFOX2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001031695.2 NP_001026865.1 12, 13 1459
          NM_001082576.1 NP_001076045.1 12, 13 1447
          NM_001082577.1 NP_001076046.1 11, 12 1419
          NM_001082578.1 NP_001076047.1 13, 14 1419
          NM_001082579.1 NP_001076048.1 13, 14 1416
          NM_014309.2 NP_055124.1 11, 12 1410
          XM_005261428.2 XP_005261485.1 14, 15 1794
          XM_005261429.2 XP_005261486.1 14, 15 1791
          XM_005261430.2 XP_005261487.1 13, 14 1765
          XM_005261431.2 XP_005261488.1 13, 14 1762
          XM_005261432.2 XP_005261489.1 13, 14 1759
          XM_005261433.2 XP_005261490.1 12, 13 1722
          XM_005261435.1 XP_005261492.1 13, 14 1495
          XM_005261437.1 XP_005261494.1 12, 13 1466
          XM_006724185.2 XP_006724248.1 14, 15 1806
          XM_006724186.2 XP_006724249.1 14, 15 1803
          XM_006724187.2 XP_006724250.1 13, 14 1777
          XM_006724188.2 XP_006724251.1 12, 13 1734
          XM_006724189.3 XP_006724252.1 14, 15 1499
          XM_006724190.1 XP_006724253.1 14, 15 1193
          XM_006724191.1 XP_006724254.1 13, 14 1498
          XM_006724192.2 XP_006724255.1 13, 14 1328
          XM_006724194.2 XP_006724257.1 13, 14 1270
          XM_017028686.1 XP_016884175.1 14, 15 1185
          XM_017028687.1 XP_016884176.1 14, 15 1366
          XM_017028688.1 XP_016884177.1 12, 13 1719
          XM_017028689.1 XP_016884178.1 14, 15 1259
          XM_017028690.1 XP_016884179.1 13, 14 1248
          XM_017028691.1 XP_016884180.1 13, 14 1245
          XM_017028692.1 XP_016884181.1 13, 14 1496
          XM_017028693.1 XP_016884182.1 13, 14 1486
          XM_017028694.1 XP_016884183.1 13, 14 1483
          XM_017028695.1 XP_016884184.1 12, 13 1171
          XM_017028696.1 XP_016884185.1 12, 13 1454
          XM_017028697.1 XP_016884186.1 14, 15 2275
          XM_017028698.1 XP_016884187.1 11, 12 1411
          GenBank mRNA AB649123.1 - 13, 14 1126
          AF229058.1 - 12, 13 1264
          AK055213.1 - 13, 14 1380
          AK091695.1 - 2, 3 2230
          AK291460.1 - 12, 13 1343
          AK314632.1 - 12, 13 1278
          AL009266.1 - 11, 12 1215
          AM419009.1 - 13, 14 1138
          AY072786.1 - 13, 14 1229
          AY960684.1 - 11, 12 1054
          BC013115.1 - 11, 12 1217 4346741
          BC025281.1 - 12, 13 1282 4555865
          CR456559.1 - 12, 13 1148
          DQ778625.1 - 13, 14 1307

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA processing factor RNA splicing factor nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of alternative mRNA splicing, via spliceosome
          mRNA processing
          nervous system development
          RNA splicing
          fibroblast growth factor receptor signaling pathway
          regulation of definitive erythrocyte differentiation
          RNA metabolic process
          radial glia guided migration of Purkinje cell
          intracellular estrogen receptor signaling pathway
          regulation of cell proliferation
          negative regulation of transcription, DNA-templated
          dendrite morphogenesis
          neuromuscular process controlling balance
          nucleotide binding
          transcription corepressor activity
          RNA binding
          mRNA binding
          protein binding
          transcription factor binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-sskjb:80/100.66.75.127:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0