Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPIL2 SIRNA Assays ›
          Gene Symbol:
          PPIL2
          Gene Name:
          peptidylprolyl isomerase like 2
          Gene Aliases:
          CYC4, CYP60, Cyp-60, UBOX7, hCyP-60
          Chromosome Location:
          Chr.22: 21665984 - 21698276 on Build GRCh38
          UniGene ID:
          Hs.438587
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS177570
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPIL2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001317996.1 NP_001304925.1 15, 16 1244
          NM_014337.3 NP_055152.1 15, 16 1244
          NM_148175.2 NP_680480.1 15, 16 1244
          NM_148176.2 NP_680481.1 15, 16 1244
          XM_005261448.3 XP_005261505.1 15, 16 1244
          XM_011530041.2 XP_011528343.1 15, 16 1244
          XM_011530042.2 XP_011528344.1 15, 16 1244
          XM_011530043.2 XP_011528345.1 15, 16 1244
          XM_011530044.2 XP_011528346.1 15, 16 1244
          XM_011530046.2 XP_011528348.1 15, 16 1244
          XM_011530047.2 XP_011528349.1 15, 16 1244
          XM_011530049.2 XP_011528351.1 16, 17 1294
          XM_011530050.2 XP_011528352.1 14, 15 1157
          XM_011530051.2 XP_011528353.1 15, 16 1184
          XM_017028705.1 XP_016884194.1 6, 7 490
          XM_017028706.1 XP_016884195.1 6, 7 490
          GenBank mRNA AK127259.1 - 17, 18 1356
          AK289545.1 - 15, 16 1206
          AK299206.1 - 14, 15 1106
          BC000022.2 - 15, 16 1206 3505936
          BC028385.2 - 15, 16 1188 4816793
          CR456548.1 - 15, 16 1160
          U37219.1 - 15, 16 1220
          U37220.1 - 6, 7 468
          U37221.1 - 6, 7 503

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          protein peptidyl-prolyl isomerization
          protein folding
          leukocyte migration
          protein localization to plasma membrane
          peptidyl-prolyl cis-trans isomerase activity
          protein binding
          ligase activity
          ubiquitin-ubiquitin ligase activity
          ubiquitin protein ligase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-wtw7d:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0