Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MTHFD1L SIRNA Assays ›
          Gene Symbol:
          MTHFD1L
          Gene Name:
          methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1-like
          Gene Aliases:
          FTHFSDC1, MTC1THFS, dJ292B18.2
          Chromosome Location:
          Chr.6: 150865679 - 151101887 on Build GRCh38
          UniGene ID:
          Hs.591343
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS177905
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MTHFD1L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242767.1 NP_001229696.1 21, 22 2413
          NM_001242768.1 NP_001229697.1 21, 22 2097
          NM_015440.4 NP_056255.2 21, 22 2410
          XM_005266907.4 XP_005266964.1 21, 22 2351
          XM_005266910.4 XP_005266967.1 21, 22 2354
          XM_011535729.2 XP_011534031.1 21, 22 2354
          XM_011535730.2 XP_011534032.1 21, 22 2097
          XM_011535731.2 XP_011534033.1 21, 22 2069
          XM_011535732.2 XP_011534034.1 21, 22 2151
          XM_011535733.2 XP_011534035.1 21, 22 2597
          XM_011535734.2 XP_011534036.1 21, 22 2190
          XM_017010702.1 XP_016866191.1 21, 22 2353
          XM_017010703.1 XP_016866192.1 21, 22 2353
          XM_017010704.1 XP_016866193.1 21, 22 2069
          XM_017010705.1 XP_016866194.1 21, 22 2594
          XM_017010706.1 XP_016866195.1 21, 22 2597
          XM_017010707.1 XP_016866196.1 21, 22 2594
          GenBank mRNA AB127387.1 - 21, 22 2266
          AK024798.1 - 5, 6 498
          AK127089.1 - 20, 21 2889
          AK315378.1 - 20, 21 2206
          AL117452.1 - 21, 22 2085
          AY374130.1 - 21, 22 2373
          BC008629.1 - 7, 8 679
          BC017477.1 - 16, 17 1657
          BC110319.1 - 20, 21 2032
          BC136685.1 - 21, 22 2303 9020675
          BC144368.1 - 21, 22 2303 9052889
          BC144369.1 - 21, 22 2300

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube closure
          one-carbon metabolic process
          folic acid-containing compound metabolic process
          folic acid-containing compound biosynthetic process
          formate metabolic process
          tetrahydrofolate interconversion
          tetrahydrofolate metabolic process
          folic acid metabolic process
          embryonic neurocranium morphogenesis
          embryonic viscerocranium morphogenesis
          oxidation-reduction process
          formate-tetrahydrofolate ligase activity
          methenyltetrahydrofolate cyclohydrolase activity
          methylenetetrahydrofolate dehydrogenase (NADP+) activity
          ATP binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0