Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: L3MBTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015478.6 | NP_056293.4 | 11 | 1365 | ||
NM_032107.4 | NP_115479.4 | 14 | 1536 | |||
GenBank mRNA | AB014581.1 | - | 7 | 615 | ||
AK299199.1 | - | 14 | 1506 | |||
AL110279.1 | - | 3 | 2057 | |||
BC039820.1 | - | 11 | 1280 | 5199689 |