Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278716.1 | NP_001265645.1 | 9 | 2196 | ||
NM_012160.4 | NP_036292.2 | 8 | 2078 | |||
NR_103836.1 | 7 | 1732 | ||||
XM_005266930.2 | XP_005266987.1 | 8 | 2112 | |||
XM_017010726.1 | XP_016866215.1 | 8 | 2010 | |||
XM_017010727.1 | XP_016866216.1 | 7 | 1966 | |||
XM_017010728.1 | XP_016866217.1 | 10 | 2272 | |||
GenBank mRNA | AF174590.1 | - | 6 | 1687 | ||
AF176699.1 | - | 6 | 1687 | |||
AF199355.1 | - | 6 | 1691 | |||
AJ420594.1 | - | 2 | 341 | |||
AK308736.1 | - | 3 | 563 | |||
AK313076.1 | - | 8 | 2007 | |||
BC055010.1 | - | 8 | 2017 | 6137889 | ||
BC091484.1 | - | 9 | 2116 | 6178874 |