Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B9D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243473.2 | NP_001230402.1 | 6 | 904 | ||
NM_001321214.1 | NP_001308143.1 | 7 | 758 | |||
NM_001321215.1 | NP_001308144.1 | 6 | 997 | |||
NM_015681.4 | NP_056496.1 | 7 | 669 | |||
XM_005256607.2 | XP_005256664.1 | 7 | 675 | |||
GenBank mRNA | AB030506.1 | - | 7 | 658 | ||
AI300844.1 | - | 1 | 265 | |||
AK293815.1 | - | 6 | 970 |