Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GK SIRNA Assays ›
          Gene Symbol:
          GK
          Gene Name:
          glycerol kinase
          Gene Aliases:
          GK1, GKD
          Chromosome Location:
          Chr.X: 30653348 - 30731462 on Build GRCh38
          UniGene ID:
          Hs.1466
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID HSS178323
          Size 20 nmol
          Availability Made To Order
          Catalog # 1299001
          Price Contact Us
          Your Price
          Online offer:
          3062.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000167.5 NP_000158.1 12 1122
          NM_001128127.2 NP_001121599.1 12 1122
          NM_001205019.1 NP_001191948.1 13 1140
          NM_203391.3 NP_976325.1 13 1140
          XM_005274488.4 XP_005274545.1 10 892
          XM_006724483.2 XP_006724546.1 13 1159
          XM_006724484.2 XP_006724547.1 13 1158
          XM_006724485.2 XP_006724548.1 12 985
          XM_006724486.3 XP_006724549.1 11 970
          XM_011545491.2 XP_011543793.1 14 1178
          XM_011545492.1 XP_011543794.1 14 1177
          XM_011545493.2 XP_011543795.1 12 860
          XM_011545494.2 XP_011543796.1 12 805
          XM_017029409.1 XP_016884898.1 14 1175
          XM_017029410.1 XP_016884899.1 11 954
          XM_017029411.1 XP_016884900.1 11 969
          XM_017029412.1 XP_016884901.1 10 890
          GenBank mRNA AK294304.1 - 13 946
          AK294938.1 - 14 1175
          AK308238.1 - 10 801
          AK313215.1 - 12 1058
          BC037549.1 - 12 979 5213716
          BC042421.1 - 13 1141 4820945
          BC071595.1 - 13 1141 5276056
          BX647604.1 - 13 961
          L13943.1 - 12 1009
          X68285.1 - 11 733
          X69886.1 - 8 558

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          carbohydrate kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          glycerol metabolic process
          triglyceride metabolic process
          response to cold
          phosphorylation
          regulation of fatty acid metabolic process
          triglyceride biosynthetic process
          glycerol catabolic process
          response to drug
          glucose homeostasis
          response to ethanol
          glycerol-3-phosphate biosynthetic process
          glycerol kinase activity
          protein binding
          ATP binding
          histone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline